missing translation for 'onlineSavingsMsg'
Learn More

BOS-172722, MedChemExpress

Product Code. 16434588
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434588

Brand: Medchem Express HY112162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BOS-172722 is an inhibitor of monopolar spindle 1 (MPS1) checkpoint with an IC50 of 11 nM and 63 nM for MPS1 (1 mM ATP) and P-MPS1, respectively. BOS-172722 also has potential for the study of various forms of breast cancer.

BOS-172722 is an inhibitor of monopolar spindle 1 (MPS1) checkpoint with an IC50 of 11 nM and 63 nM for MPS1 (1 mM ATP) and P-MPS1, respectively. BOS-172722 also has potential for the study of various forms of breast cancer.

Specifications

Chemical Name or Material BOS-172722
CAS 1578245-44-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H30N8O
Quantity 10 mg
Solubility Information 1M HCl : 65 mg/mL (145.56 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 5 mg/mL (11.20 mM; Need ultrasonic)
SMILES CC1=CC2=CN=C(NC3=CC=C(C4=NN=CN4C)C=C3OCC)N=C2C(NCC(C)(C)C)=N1
Molecular Weight (g/mol) 446.55
Formula Weight 446.55
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.