Learn More
Witeg™ Borosilicate, Graduated Cylinders, Thread & Screw Cap
Product Code. 10670853
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
Subdivision:
1mL
2mL
Unit Size:
Each
Pack of 10
Product Code. 10670853
Brand: Witeg™ 3 751 010
Only null left
Add to basket
Add to basket
Measure the volume of liquids in the lab with the Witeg™ Borosilicate, Graduated Cylinders, Thread & Screw Cap.
  • Glass
  • Graduation
  • Thread
  • Screw Cap
  • Glass
  • Graduation
  • Thread
  • Screw Cap
Specifications
Base Type Hexagonal
Scale Color Brown
Shape Round
Certifications/Compliance DIN EN ISO 4788
Class B
Tolerance Metric ±1 mL
Type Graduated Cylinder
Material Borosilicate Glass
Capacity (Metric) 100 mL
Includes Screw Cap
Graduations 2 mL
Subdivision 2mL