Learn More
Karl Hecht™ Assistent™ Borosilicate Round Bottom Flasks
Can be used to make solutions or hold, contain, collect and measure chemicals. Karl Hecht™ Assistent™ Borosilicate Round Bottom Flasks are made from durable DURAN™ glass 3.3.
Brand:  Karl Hecht™ 41920080
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10709894
  • 33.71 € / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Joint size durably marked
  • Without stopper
  • With standard ground, reinforced neck
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Stopper Number: NS 29/32
1000 mL
Borosilicate Glass