Learn More
Karl Hecht™ Assistent™ Borosilicate Round Bottom Flasks
Can be used to make solutions or hold, contain, collect and measure chemicals. Karl Hecht™ Assistent™ Borosilicate Round Bottom Flasks are made from durable DURAN™ glass 3.3.
Brand: Karl Hecht™ 41920020
Additional Details : Weight : 0.10000kg
Description
- Joint size durably marked
- Without stopper
- With standard ground, reinforced neck
- Corrosion-free and resistant to thermal shock
Specifications
Stopper Number: NS 14/23 | |
25 mL | |
Borosilicate Glass |