missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Hecht™ Erlenmeyer Flasks, Graduated, Narrow Neck
Conical, flat-bottomed flask well suited to mixing by swirling. Karl Hecht™ Erlenmeyer Flask, Graduated, Narrow Neck is constructed of durable borosilicate 3.3 glass.
Brand: Karl Hecht™ 41940154
Additional Details : Weight : 0.10000kg
Description
- Durable borosilicate 3.3 glass.
- Graduated.
- Narrow neck.
Specifications
DIN ISO 1773 | |
42 mm | |
Borosilicate Glass (3.3) | |
Narrow |
131 mm | |
1000 mL | |
220 mm | |
Yes |