Learn More
Robu™ Borosilicate Glass Filter Funnels
Brand:  Robu™ 22254
Additional Details : Weight : 0.33720kg
 View more versions of this product
Product Code. 16843046
  • 282.00 € / Pack of 10
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Specifications
55mm (OD); 25mm (ID)
8mm
Use in sample preparation processes and the filtration of small amounts
4
25 mL
Borosilicate Glass
55mm
Filter funnel