Learn More
Robu™ Borosilicate Glass Filter Funnels
Brand:  Robu™ 22253
Additional Details : Weight : 0.33720kg
 View more versions of this product
Product Code. 10193831
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
55mm (OD); 25mm (ID)
8mm
Use in sample preparation processes and the filtration of small amounts
3
25 mL
Borosilicate Glass
55mm
Filter funnel