Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Filter Funnels
Lenz Laborglasinstrumente™ Filter funnels are ideal for separating solids from liquids via laboratory filtration.
Brand:  Lenz Laborglasinstrumente™ 07195340
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10767331
  • 75.05 € / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Made from borosilicate glass
  • Separates solids from liquids
Specifications
Specifications
125 mL
140 mm
3
Borosilicate Glass
60mm
Filter funnel