Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Filter Funnels
Lenz Laborglasinstrumente™ Filter funnels are ideal for separating solids from liquids via laboratory filtration.
Brand:  Lenz Laborglasinstrumente™ 07195458
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10460133
  • 146.28 € / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Made from borosilicate glass
  • Separates solids from liquids
Specifications
Specifications
500 mL
200 mm
4
Borosilicate Glass
95mm
Filter funnel