Learn More
Pyrex™ Borosilicate Glass Distillation Flasks
Complies with many ASTM test method requirements. Pyrex™ Borosilicate Glass Distillation Flasks require use of a calibrated thermomenter for 76mm immersion.
Brand: Pyrex™ 1290/06M
Description
- Complies with ASTM E133 and E1405
- For the distillation of petroleum products
- 1290/04M complies with the requirements for ASTM test methods D86, D233, D447, D801, D802
- 1290/06M complies with the requirements for ASTM test method D447
- Side arm inclined at 75° to the vertical axis
- For use with thermometers calibrated for 76mm immersion
- Length of side arm is 100mm