Learn More
Pyrex™ Borosilicate Glass Distillation Flasks
Complies with many ASTM test method requirements. Pyrex™ Borosilicate Glass Distillation Flasks require use of a calibrated thermomenter for 76mm immersion.
Brand:  Pyrex™ 1290/06M
Product Code. 11317844
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Complies with ASTM E133 and E1405
  • For the distillation of petroleum products
  • 1290/04M complies with the requirements for ASTM test methods D86, D233, D447, D801, D802
  • 1290/06M complies with the requirements for ASTM test method D447
  • Side arm inclined at 75° to the vertical axis
  • For use with thermometers calibrated for 76mm immersion
  • Length of side arm is 100mm