missing translation for 'onlineSavingsMsg'
Learn More

Hecht Karl™ Borosilicate Glass Crystallizing Dish With Spout

Brand:  Hecht Karl™ 41969040

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10757502

  • 6.31 € / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Flat
Round
25 mm
Borosilicate Glass
Crystallizing Dish
40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions