Learn More
MedChemExpress Bombesin
Specifications
Specifications
| Purity | 99.76% |
| CAS | 31362-50-2 |
| Content And Storage | Protect from light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| Molecular Weight (g/mol) | 1619.85 g/mol |
| Product Type | Natural Products |
| Quantity | 5 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 1.43 mg/mL (0.88 mM∣Need ultrasonic) |