Learn More
Boldenone Undecanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Boldenone Undecanoate |
| CAS | 7219-21-8 |
| Molecular Formula | C30 H46 O3 |
| Quantity | 250 mg |
| Synonym | (17beta)-17-[(1-Oxoundecyl)oxy] Androsta-1,4-dien-3-one, 17beta-Hydroxy-a ndrosta-1,4-dien-3-one Undecanoate |
| InChI Formula | InChI=1S/C30H46O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h17,19,21,24-27H,4-16,18,20H2,1-3H3/t24-,25-,26-,27-,29-,30-/m0/s1 |
| SMILES | CCCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] undecanoate |
| Molecular Weight (g/mol) | 454.684 |
| Formula Weight | 454.345 |