Learn More
Bohlender™ BOLA™ UNF Screw-in Tube Fittings
Product Code. 15546965
Change view
Click to view available options
Diameter (English) Inner:
0.03 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Unit Size
15546965 0.03 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15546965 Supplier Bohlender™ Supplier No. F76308
  • ONLINE EXCLUSIVE PRICE
    52.50 €
    42.19 € / Pack of 1
    Save 10.31 €
    19% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
UNF screw-in tube fittings made of PTFE.
  • Straight tube fitting made of PTFE
  • One side with fitting thread GL 14 for connecting hard-walled tubes and tubing by means of BOLA™ Laboratory Screw Joints
  • Other side with male screw-in thread UNF 0.25 in. 28G for connection to units and fittings with female thread UNF 0.25 in. 28G
  • Bore diameter: 0.8 mm
Specifications
Diameter (English) Inner 0.03 in.
Diameter (Metric) Inner 0.8 mm
Material PTFE
Diameter (English) Outer 0.06 in.
Diameter (Metric) Outer 1.6 mm
Overall Length (English) 1.5 in.
Overall Length (Metric) 39 mm
For Use With BOLA™ Laboratory Screw Joints
Thread Size GL 14, Male thread UNF 0.25 in. 28G