Learn More
Bohlender™ BOLA™ Measuring Cylinders
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
Graduations:
1 mL
2 mL
2.5 mL
5 mL
Diameter (Metric) Inside:
24 mm
30 mm
40 mm
53 mm
Height (Metric):
172 mm
202 mm
268 mm
308 mm
Unit Size:
Each
Description
- Translucent and non-porous
- Design based on ISO 4788
- With graduation (uncalibrated), spout and reinforcing ring
- Universal chemical resistance
- Product is only exposed to PTFE
- Hexagonal base prevents the cylinder from rolling
- Permanent, recessed graduation
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | PTFE |
| Graduations | 5 mL |
| Capacity (English) | 17 oz. |
| Shape | Cylinder |
| Diameter (Metric) Inside | 53 mm |
| Diameter (English) Inside | 2.1 in. |
| Height (English) | 12.1 in. |
| Height (Metric) | 308 mm |