missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ GLS Reductions, PTFE and PP
Description
- Blue screw cap made of PP with GLS 80 thread
- Movable insert with GL 45 thread made of PTFE
- Transition from GLS 80 bottles to a GL 45 thread
- Body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fixed on another vessel without the risk of disarranging the tubing or cable
- Product is only exposed to PTFE
Specifications
Specifications
| Material | Polypropylene, PTFE |
| For Use With (Application) | Transition From GLS 80 Bottles to GL 45 Thread |
| Thread Size | GLS 80 (Reducing Thread GL 45) |
| Color | Blue |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?