missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ GLS Reductions, PP
Description
- Blue screw cap made of PP
- Movable reduction body made of PP
- O-ring made of FPM for transition to GL threads
- Body can be moved independently from the screw cap so that the completely assembled reduction can be removed and fixed on another vessel without the risk of disarranging the tubing or cable
Specifications
Specifications
| Material | Polypropylene |
| For Use With (Application) | .8 to 8 mm ID Tubing |
| Thread Size | GLS 80 (Reducing Thread GL 14) |
| Color | Blue |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?