Learn More
Bohlender™ BOLA™ Gas Inlet Tubes
Product Code. 17874034
Change view
Click to view available options
Quantity:
5 Pk.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17874034 5 Pk. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17874034 Supplier Bohlender™ Supplier No. N91101
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Gas Inlet Tubes made of FEP for Scrubber Adapters for Bottles and Scrubber Bottles Vitrum.
  • Suitable for BOLA™ Scrubber Adaptors GL 45 Scrubber Bottles Vitrum GL 45, Cat. No. N 1660-14, N 1662-14, N 1662-24
  • Suitable for BOLA™ Scrubber Adaptors GLS 80 Scrubber Bottles Vitrum GL 45, Cat. No. N 1660-24, N 1662-34, N 1662-44
Specifications
Material FEP
For Use With (Equipment) BOLA™ Scrubber Adapters for Bottles and BOLA™ Scrubber Bottles Vitrum
Quantity 5 Pk.