missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boekel Scientific™ TropiCooler™ Incubator Blocks
Ideal for restriction enzymes and DNA libraries
Brand: Boekel Scientific™ 260052
Additional Details : Weight : 2.00000kg
Description
- Machined aluminum blocks with anodized finish and self-contained recessed handle
- Hold 0.5 and 1.5mL microcentrifuge tubes (13-245-114 holds seven of each tube size) Dimensions, 21/8L × 31/4W × 11/2 in.H (5.4 × 8.3 × 3.8cm)
Specifications
Boekel TropiCooler incubator | |
Aluminum | |
For 0.5mL microcentrifuge tube |
0.5mL | |
Accessory, Block with Extractor |