Learn More
Boekel Scientific™ Platform for Microprocessor Controlled Orbital Shaker
Click to view available options
Holds:
12 x 100 mL Flasks, 12 x 150 mL Beakers
6 x 250 mL Flasks, 6 x 400 mL Beakers
Unit Size:
Each
Description
- Platform with Holders for Flasks and Beakers
- interchangeable Locking Design
Specifications
Specifications
| Load Bearing Capacity (Metric) | 2.5 kg |
| Holds | 6 x 250 mL Flasks, 6 x 400 mL Beakers |
| Product Type | Platform |
| Length (English) | 9.8 in. |
| Length (Metric) | 250 mm |
| Width (English) | 7.5 in. |
| Width (Metric) | 190 mm |
| For Use With (Equipment) | Boekel Microprocessor Controlled Orbital Shaker |