Learn More
Boekel Scientific™ Platform for Microprocessor Controlled Orbital Shaker
GREENER_CHOICE
Product Code. 12693555
Click to view available options
Holds:
Plates
Unit Size:
Each
Product Code. 12693555
Brand: Boekel Scientific™ 270293
Only null left
Add to basket
Add to basket
This is an interchangeable platform for Boekel Orbital Shakers (Cat. No. 15-600-317).
  • Flat Platform with Rubber Mat
  • Nonslip Design for All Applications
  • interchangeable Locking Design
Specifications
Load Bearing Capacity (Metric) 2.5 kg
Holds Plates
Product Type Flat Platform with Rubber Mat
Depth (English) 8.7 in.
Depth (Metric) 22 cm
Length (English) 8.7 in.
Length (Metric) 22 cm
Width (English) 8.7 in.
Width (Metric) 220 mm
For Use With (Equipment) Boekel Microprocessor Controlled Orbital Shaker