Learn More
Boekel Scientific™ Microplate Shaker Platform
GREENER_CHOICE
Product Code. 12613565
Click to view available options
Holds:
4 Microplates
Unit Size:
Each
Product Code. 12613565
Brand: Boekel Scientific™ 270390
Please to purchase this item. Need a web account? Register with us today!
Need to upgrade your Boekel Microplate Shaker to hold more plates? This 4 plate microplate tray is adaptable to the Boekel 2 Plate Microplate Shaker.
  • interchangeable tray for holding microplates
  • Simple screw attachments allow quick and easy fitting of any standard depth 96- or 384-well plates, holding the plates securely in place
Specifications
Holds 4 Microplates
For Use With (Equipment) Boekel Scientific Microplate Shaker
No. of Positions 4