Learn More
Boekel Scientific™ Microplate Shaker Platform
Click to view available options
Holds:
4 Microplates
Unit Size:
Each
Description
- interchangeable tray for holding microplates
- Simple screw attachments allow quick and easy fitting of any standard depth 96- or 384-well plates, holding the plates securely in place
Specifications
Specifications
| Holds | 4 Microplates |
| For Use With (Equipment) | Boekel Scientific Microplate Shaker |
| No. of Positions | 4 |