missing translation for 'onlineSavingsMsg'
Learn More

Bode Chemie™ Bodedex Forte Instrument Cleaner

Product Code. 10324321
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
10324321 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10324321 Supplier Bode Chemie™ Supplier No. 980250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Instrument Cleaner
Form Liquid
Appearance Pale Yellow
Color Yellow
Composition Non-ionic and Amphoteric Surfactants, solvents, complexing agents, amino acid derivatives, corrosion
Container Type Canister
For Use With (Application) Heat-Sensitive And Heat-Resistant Instruments
Volume (Metric) 5000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.