missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Boc-Gly-Gly-Phe-Gly-OH TFA
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Purity | 98.27% |
| Content And Storage | Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 550.48 g/mol |
| Product Type | Linker |
| Quantity | 10 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 125 mg/mL (227.07 mM∣Need ultrasonic) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction