Learn More
Boc-2-iodo-L-phenylalanine, TRC
Specifications
Specifications
| Chemical Name or Material | Boc-2-iodo-L-phenylalanine |
| CAS | 273221-78-6 |
| Molecular Formula | C14 H18 I N O4 |
| Quantity | 1 g |
| Synonym | N-[(1,1-Dimethylethoxy)carbonyl]-2-iodo-L-phenylalanine |
| InChI Formula | InChI=1S/C14H18INO4/c1-14(2,3)20-13(19)16-11(12(17)18)8-9-6-4-5-7-10(9)15/h4-7,11H,8H2,1-3H3,(H,16,19)(H,17,18)/t11-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1I)C(=O)O |
| IUPAC Name | (2S)-3-(2-iodophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| Molecular Weight (g/mol) | 391.202 |
| Formula Weight | 391.028 |