Learn More
Bobcat339 hydrochloride, MedChemExpress
Product Code. 16447217
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16447217
Brand: Medchem Express HY111558A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Bobcat339 hydrochloride is a potent and selective cytosine-based inhibitor of TET enzyme, with the IC50s of 33 μM and 73 μM for TET1 and TET2, respectively. Bobcat339 hydrochloride is useful to the field of epigenetics and serves as a starting point for new therapeutics that target DNA methylation and gene transcription.
Bobcat339 hydrochloride is a potent and selective cytosine-based inhibitor of TET enzyme, with the IC50s of 33 μM and 73 μM for TET1 and TET2, respectively. Bobcat339 hydrochloride is useful to the field of epigenetics and serves as a starting point for new therapeutics that target DNA methylation and gene transcription.
Specifications
Chemical Name or Material Bobcat339 hydrochloride
Physical Form Solid
CAS 2436747-44-1
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C16H13Cl2N3O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 130 mg/mL (388.99 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.30 mM; Need ultrasonic)
SMILES NC1=NC(N(C2=CC=CC(C3=CC=CC=C3)=C2)C=C1Cl)=O.Cl
Molecular Weight (g/mol) 334.2
Formula Weight 334.2
Percent Purity 98.08%
Grade Research