missing translation for 'onlineSavingsMsg'
Learn More

Bobcat339 hydrochloride, MedChemExpress

Product Code. 16424498
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424498

Brand: Medchem Express HY111558A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bobcat339 hydrochloride is a potent and selective cytosine-based inhibitor of TET enzyme, with the IC50s of 33 μM and 73 μM for TET1 and TET2, respectively. Bobcat339 hydrochloride is useful to the field of epigenetics and serves as a starting point for new therapeutics that target DNA methylation and gene transcription.

Bobcat339 hydrochloride is a potent and selective cytosine-based inhibitor of TET enzyme, with the IC50s of 33 μM and 73 μM for TET1 and TET2, respectively. Bobcat339 hydrochloride is useful to the field of epigenetics and serves as a starting point for new therapeutics that target DNA methylation and gene transcription.

Specifications

Chemical Name or Material Bobcat339 hydrochloride
Physical Form Solid
CAS 2436747-44-1
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H13Cl2N3O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 130 mg/mL (388.99 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.30 mM; Need ultrasonic)
SMILES NC1=NC(N(C2=CC=CC(C3=CC=CC=C3)=C2)C=C1Cl)=O.Cl
Molecular Weight (g/mol) 334.2
Formula Weight 334.2
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.