missing translation for 'onlineSavingsMsg'
Learn More

BMX-IN-1, MedChemExpress

Product Code. 16400698
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16400698

Brand: Medchem Express HY80002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMX-IN-1 is a selective, irreversible inhibitor of bone marrow tyrosine kinase on chromosome X (BMX) that targets Cys496 in the BMX ATP binding domain with an IC50 of 8 nM, also targets the related Bruton’s tyrosine kinase (BTK) with an IC50 value of 10.4 nM, but is more than 47-656-fold less potent against Blk, JAK3, EGFR, Itk, or Tec activity.

BMX-IN-1 is a selective, irreversible inhibitor of bone marrow tyrosine kinase on chromosome X (BMX) that targets Cys496 in the BMX ATP binding domain with an IC50 of 8 nM, also targets the related Bruton’s tyrosine kinase (BTK) with an IC50 value of 10.4 nM, but is more than 47-656-fold less potent against Blk, JAK3, EGFR, Itk, or Tec activity.

Specifications

Chemical Name or Material BMX-IN-1
CAS 1431525-23-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H24N4O4S
Quantity 5 mg
Synonym BMX kinase inhibitor
Solubility Information DMF : 10 mg/mL (19.06 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (15.88 mM; Need ultrasonic)
SMILES O=C1N(C2=C(C=C1)C=NC3=CC=C(C4=CC=C(NS(=O)(C)=O)C=C4)C=C32)C5=CC=C(C)C(NC(C=C)=O)=C5
Molecular Weight (g/mol) 524.59
Formula Weight 524.59
Percent Purity 96.27%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.