Learn More
BMS-986020, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 1-(4'-(3-Methyl-4-(((((R)-1-phenylethyl)oxy)carbonyl)amino)isoxazol-5-yl)biphenyl-4-yl)cyclopropanecarboxylic Acid |
| CAS | 1257213-50-5 |
| Molecular Formula | C29H26N2O5 |
| Quantity | 50 mg |
| Synonym | BMS-986020, 1-[4'-[3-Methyl-4-[[[((R)-1-phenylethyl)oxy]carbonyl]amino]isoxazol-5-yl]biphenyl-4-yl]cyclopropanecarboxylic Acid |
| InChI Formula | InChI=1S/C29H26N2O5/c1-18-25(30-28(34)35-19(2)20-6-4-3-5-7-20)26(36-31-18)23-10-8-21(9-11-23)22-12-14-24(15-13-22)29(16-17-29)27(32)33/h3-15,19H,16-17H2,1-2H3,(H,30,34)(H,32,33)/t19-/m1/s1 |
| SMILES | C[C@@H](OC(=O)Nc1c(C)noc1c2ccc(cc2)c3ccc(cc3)C4(CC4)C(=O)O)c5ccccc5 |
| IUPAC Name | 1-[4-[4-[3-methyl-4-[[(1R)-1-phenylethoxy]carbonylamino]-1,2-oxazol-5-yl]phenyl]phenyl]cyclopropane-1-carboxylic acid |
| Molecular Weight (g/mol) | 482.53 |
| Formula Weight | 482.1842 |