missing translation for 'onlineSavingsMsg'
Learn More

BMS-986020, MedChemExpress

Product Code. 16494396
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Código de producto. 16494396

missing translation for 'mfr': Medchem Express HY100619

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

MedChemExpress BMS-986020 (AM152) is a high-affinity and selective lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 inhibits bile acid and phospholipid transporters with IC50s of 4.8 μM, 6.2 μM, and 7.5 μM for BSEP, MRP4, and MDR3, respectively. BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF).

BMS-986020 (AM152) is a high-affinity and selective lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 inhibits bile acid and phospholipid transporters with IC50s of 4.8 μM, 6.2 μM, and 7.5 μM for BSEP, MRP4, and MDR3, respectively. BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF).

Specifications

Chemical Name or Material BMS-986020
Physical Form Solid
CAS 1257213-50-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C29H26N2O5
For Use With (Application) COVID-19-immunoregulation
Synonym AM152
Solubility Information DMSO : 250 mg/mL (518.10 mM; Need ultrasonic)
SMILES O=C(C1(C2=CC=C(C3=CC=C(C4=C(NC(O[C@@H](C5=CC=CC=C5)C)=O)C(C)=NO4)C=C3)C=C2)CC1)O
Molecular Weight (g/mol) 482.53
Formula Weight 482.53
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.