Learn More
BMS-986020, MedChemExpress
Product Code. 16499656
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16499656
Brand: Medchem Express HY100619
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress BMS-986020 (AM152) is a high-affinity and selective lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 inhibits bile acid and phospholipid transporters with IC50s of 4.8 μM, 6.2 μM, and 7.5 μM for BSEP, MRP4, and MDR3, respectively. BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF).
BMS-986020 (AM152) is a high-affinity and selective lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 inhibits bile acid and phospholipid transporters with IC50s of 4.8 μM, 6.2 μM, and 7.5 μM for BSEP, MRP4, and MDR3, respectively. BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF).
Specifications
Chemical Name or Material BMS-986020
Physical Form Solid
CAS 1257213-50-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C29H26N2O5
For Use With (Application) COVID-19-immunoregulation
Synonym AM152
Solubility Information DMSO : 250 mg/mL (518.10 mM; Need ultrasonic)
SMILES O=C(C1(C2=CC=C(C3=CC=C(C4=C(NC(O[C@@H](C5=CC=CC=C5)C)=O)C(C)=NO4)C=C3)C=C2)CC1)O
Molecular Weight (g/mol) 482.53
Formula Weight 482.53
Percent Purity 98.0%
Grade Research