missing translation for 'onlineSavingsMsg'
Learn More

BMS 777607, MedChemExpress

Product Code. p-200098233
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497707 5 mg 5mg
16465746 100 mg 100mg
16454978 10 mg 10mg
16421227 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497707 Supplier Medchem Express Supplier No. HY12076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases.

BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases.

Specifications

Chemical Name or Material BMS 777607
CAS 1025720-94-8
Color White
Purity Grade Notes Research
Molecular Formula C25H19ClF2N4O4
Quantity 5 mg
Synonym BMS 817378
Solubility Information DMSO : ≥ 39 mg/mL (76.04 mM)
SMILES O=C(NC1=CC=C(C(F)=C1)OC2=C(C(N)=NC=C2)Cl)C3=C(C=CN(C3=O)C4=CC=C(C=C4)F)OCC
Molecular Weight (g/mol) 512.89
Formula Weight 512.89
Percent Purity 98.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.