missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS 777607, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases.
Specifications
Specifications
| Chemical Name or Material | BMS 777607 |
| CAS | 1025720-94-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H19ClF2N4O4 |
| Quantity | 50 mg |
| Synonym | BMS 817378 |
| Solubility Information | DMSO : ≥ 39 mg/mL (76.04 mM) |
| SMILES | O=C(NC1=CC=C(C(F)=C1)OC2=C(C(N)=NC=C2)Cl)C3=C(C=CN(C3=O)C4=CC=C(C=C4)F)OCC |
| Molecular Weight (g/mol) | 512.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction