Learn More
BMS-687453, MedChemExpress
Product Code. 16415066
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16415066
Brand: Medchem Express HY10678
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress BMS-687453 is a potent and selective PPARα agonist, with an EC50 and IC50 of 10 nM and 260 nM for human PPARα and 4100 nM and >15000 nM for PPARγ in PPAR-GAL4 transactivation assays.
BMS-687453 is a potent and selective PPARα agonist, with an EC50 and IC50 of 10 nM and 260 nM for human PPARα and 4100 nM and >15000 nM for PPARγ in PPAR-GAL4 transactivation assays.
Specifications
Chemical Name or Material BMS-687453
CAS 1000998-59-3
Color White
Purity Grade Notes Research
Molecular Formula C22H21ClN2O6
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (224.79 mM)
SMILES ClC(C=C1)=CC=C1C2=NC(COC3=CC=CC(CN(C(OC)=O)CC(O)=O)=C3)=C(C)O2
Molecular Weight (g/mol) 444.86
Formula Weight 444.86
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus