missing translation for 'onlineSavingsMsg'
Learn More

BMS-582949 hydrochloride, MedChemExpress

Product Code. 16436446
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436446

Brand: Medchem Express HY14305A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease.

BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease.

Specifications

Chemical Name or Material BMS-582949 hydrochloride
CAS 912806-16-7
Color White
Purity Grade Notes Research
Molecular Formula C22H27ClN6O2
Quantity 100 mg
Solubility Information DMSO : 25 mg/mL (56.44 mM; Need ultrasonic)
SMILES O=C(NCCC)C1=CN(N=CN=C2NC3=CC(C(NC4CC4)=O)=CC=C3C)C2=C1C.[H]Cl
Molecular Weight (g/mol) 442.94
Formula Weight 442.94
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.