Learn More
BMS-378806, MedChemExpress
Product Code. 16446396
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16446396
Brand: Medchem Express HY14134
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress BMS-378806 is a potent HIV-1 attachment inhibitor that interferes with CD4-gp120 interactions. BMS-378806 selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0.85-26.5 nM in virus.
BMS-378806 is a potent HIV-1 attachment inhibitor that interferes with CD4-gp120 interactions. BMS-378806 selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0.85-26.5 nM in virus.
Specifications
Chemical Name or Material BMS-378806
Physical Form Solid
CAS 357263-13-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H22N4O4
For Use With (Application) COVID-19-anti-virus
Synonym BMS-806
Solubility Information DMSO : 16.67 mg/mL (41.02 mM; Need ultrasonic)
SMILES COC1=C2C(NC=C2C(C(N3CCN(C[C@H]3C)C(C4=CC=CC=C4)=O)=O)=O)=NC=C1
Molecular Weight (g/mol) 406.43
Formula Weight 406.43
Percent Purity 98.61%
Grade Research