missing translation for 'onlineSavingsMsg'
Learn More

BMS-378806, MedChemExpress

Product Code. 16412117
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16412117

Brand: Medchem Express HY14134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-378806 is a potent HIV-1 attachment inhibitor that interferes with CD4-gp120 interactions. BMS-378806 selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0.85-26.5 nM in virus.

BMS-378806 is a potent HIV-1 attachment inhibitor that interferes with CD4-gp120 interactions. BMS-378806 selectively inhibits the binding of HIV-1 gp120 to the CD4 receptor with EC50 of 0.85-26.5 nM in virus.

Specifications

Chemical Name or Material BMS-378806
Physical Form Solid
CAS 357263-13-9
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H22N4O4
For Use With (Application) COVID-19-anti-virus
Synonym BMS-806
Solubility Information DMSO : 16.67 mg/mL (41.02 mM; Need ultrasonic)
SMILES COC1=C2C(NC=C2C(C(N3CCN(C[C@H]3C)C(C4=CC=CC=C4)=O)=O)=O)=NC=C1
Molecular Weight (g/mol) 406.43
Formula Weight 406.43
Percent Purity 98.61%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.