missing translation for 'onlineSavingsMsg'
Learn More

BMS-1166, MedChemExpress

Product Code. 16432826
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432826

Brand: Medchem Express HY102011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-1166 is a potent PD-1/PD-L1 immune checkpoint inhibitor. BMS-1166 induces dimerization of PD-L1 and blocks its interaction with PD-1, with an IC50 of 1.4 nM. BMS-1166 antagonizes the inhibitory effect of PD-1/PD-L1 immune checkpoint on T cell activation.

BMS-1166 is a potent PD-1/PD-L1 immune checkpoint inhibitor. BMS-1166 induces dimerization of PD-L1 and blocks its interaction with PD-1, with an IC50 of 1.4 nM. BMS-1166 antagonizes the inhibitory effect of PD-1/PD-L1 immune checkpoint on T cell activation.

Specifications

Chemical Name or Material BMS-1166
CAS 1818314-88-3
Color White
Purity Grade Notes Research
Molecular Formula C36H33ClN2O7
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (194.97 mM; Need ultrasonic)
SMILES O=C(O)[C@@H]1N(CC2=CC(Cl)=C(OCC3=CC=CC(C4=CC=C(OCCO5)C5=C4)=C3C)C=C2OCC6=CC=CC(C#N)=C6)C[C@H](O)C1
Molecular Weight (g/mol) 641.11
Formula Weight 641.11
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.