Learn More
BMP4 Monoclonal Antibody (OTI1C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803535S
Description
BMP4 Monoclonal antibody specifically detects BMP4 in Human samples. It is validated for Western BlotSpecifications
| BMP4 | |
| Monoclonal | |
| 0.5 to 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H1K1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23479 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1C2 | |
| Unconjugated | |
| ISCU | |
| BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME | |
| Full length human recombinant protein of human ISCU produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.