missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP-8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30534365
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534365 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534365 Supplier Invitrogen™ Supplier No. PA5148471

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, the BMP encoded by this gene has a proposed role in early development. In addition, the fact that this BMP is closely related to BMP5 and BMP7 has led to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene BMP8B
Gene Accession No. P34820, Q7Z5Y6
Gene Alias BMP; Bmp7r1; BMP8; BMP-8; BMP8A; BMP-8A; BMP8B; BMP-8B; Bone morphogenetic protein; bone morphogenetic protein 8 (osteogenic protein 2); bone morphogenetic protein 8A; Bone morphogenetic protein 8B; dJ118J21.1 (bone morphogenetic protein 8 (osteogenic protein 2)); MGC131757; OP2; OP-2; Op3; osteogenic protein 2
Gene Symbols BMP8A, BMP8B
Host Species Goat
Immunogen E. coli-derived recombinant human BMP-8a, Ala264-His402.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353500, 656
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.