Learn More
BML-210, MedChemExpress
Product Code. 16463717
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16463717
Brand: Medchem Express HY19350
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress BML-210 is a novel HDAC inhibitor, and its mechanism of action has not been characterized.
BML-210 is a novel HDAC inhibitor, and its mechanism of action has not been characterized.
Specifications
Chemical Name or Material BML-210
Physical Form Solid
CAS 537034-17-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H25N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 30 mg/mL (88.38 mM)
SMILES O=C(NC1=CC=CC=C1N)CCCCCCC(NC2=CC=CC=C2)=O
Molecular Weight (g/mol) 339.43
Formula Weight 339.43
Percent Purity 96.38%
Grade Research