missing translation for 'onlineSavingsMsg'
Learn More

BML-210, MedChemExpress

Product Code. 16427748
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427748

Brand: Medchem Express HY19350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BML-210 is a novel HDAC inhibitor, and its mechanism of action has not been characterized.

BML-210 is a novel HDAC inhibitor, and its mechanism of action has not been characterized.

Specifications

Chemical Name or Material BML-210
Physical Form Solid
CAS 537034-17-6
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H25N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 30 mg/mL (88.38 mM)
SMILES O=C(NC1=CC=CC=C1N)CCCCCCC(NC2=CC=CC=C2)=O
Molecular Weight (g/mol) 339.43
Formula Weight 339.43
Percent Purity 96.38%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.