Learn More
Kautex™ Blue Polypropylene Screw Closures
Product Code. 10638881
Click to view available options
For Use With (Equipment):
10 to 1000 mL Bottle
2500 mL Bottle
Diameter (Metric):
32 mm
45 mm
Unit Size:
Each
Pack of 100
Product Code. 10638881
Brand: Kautex™ 2000083951
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides tamper evidence with pull-off ring and conical seal. Kautex™ Blue Polypropylene Screw Closures are ribbed for easy grip opening.
  • Leakproof conical seal
  • Blue color for content identification
  • Polypropylene construction for strength and durability
  • Available for bottles with capacities of 10 to 2500 mL
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Includes Pull-off Ring and Conical Seal
Material Polypropylene
Thread Size (Metric) 00 mm
Neck Finish 00 mm
Product Type Screw Cap
For Use With (Equipment) 10 to 1000 mL Bottle
Diameter (Metric) 32 mm