missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Blood Group A1, A2, A3 Antigen Monoclonal Antibody (Z2B-1)

Product Code. 11505362
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11505362 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11505362 Supplier Invitrogen™ Supplier No. MA17665

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-7665 detects A1, A2, A3 human blood protein from human samples. MA1-7665 has been successfully used in agglutination applications. The MA1-7665 immunogen is A1, A2, A3 human blood protein.

Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Blood Group A1, A2, A3 Antigen
Applications Agglutination
Classification Monoclonal
Clone Z2B-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1.25mM EDTA, 0.3M NaCl and 0.1% sodium azide; pH 7.2
Gene Alias Blood Group A
Host Species Mouse
Immunogen A1, A2, A3 human blood protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.