Learn More
BD Safety-Lok™ Blood Collection Set
Use for blood collection.  BD Vacutainer™ Safety-Lok™ Blood Collection Set (50 sets) comes with safety re-shielding cover and Luer adapter.
Brand:  BD 367282
Additional Details : Weight : 0.99000kg
Product Code. 10438244
  • 93.35 € / Pack of 50
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • Includes safety re-shielding cover and Luer adapter,
  • 21 G ¾ in., 18 cm tubing, green
  • Recommended for adults
  • One case consists fo 4 x 50 Blood Collection Sets
  • One Set is necessary for a pair of blood culture bottles
Specifications
Specifications
Blood Collection Set
Sterile