Learn More
Thermo Scientific Blood Bag Adapter
Product Code. 10215893 Shop All Thermo Scientific Products
Click to view available options
Capacity:
500 mL
Unit Size:
Pack of 6
Product Code. 10215893
Brand: Thermo Scientific 76007677
Please to purchase this item. Need a web account? Register with us today!
Blood Bag Adapter for 500 mL quint, quad, triple blood bag XXL (set of 6) for double blood bucket 75007617.
  • Adapter for 500 mL quint, quad, triple blood bag ''XXL'' (set of 6) for double blood bucket 75007617.
TRUSTED_SUSTAINABILITY
Specifications
Type Blood Bag Adapter
Material Plastic
Capacity 500 mL
Autoclavable Not Autoclavable
For Use With (Equipment) Quad, Triple Blood Bag ''XXL'' (set of 6), Double Blood Bucket