Learn More
Kautex™ Black Polypropylene Screw Closures
Product Code. 15941411
Click to view available options
For Use With (Equipment):
100 mL Bottle
1500/2500/4000 mL Bottle
200/300 mL Bottle
2000 mL Bottle
250/350 mL Bottle
50 mL Bottle
500 mL Bottle
500/750/1000 mL Bottle
750/1000 mL Bottle
Diameter (Metric):
25 mm
32 mm
40 mm
45 mm
50 mm
54 mm
65 mm
80 mm
Includes:
w/Tamper Evident Ring
w/out Tamper Evident Ring
missing translation for 'unitSize'
Each
Pack of 100
Product Code. 15941411
missing translation for 'mfr': Kautex™ 2000070752
Only null left
Add to basket
Edge
Add to basket
Specifications
Color Black
Includes w/out Tamper Evident Ring
Material Polypropylene
Thread Size (Metric) 00 mm
Liner Material Polyethylene Foam Insert
Neck Finish 00 mm
Product Type Screw Cap
For Use With (Equipment) 200/300 mL Bottle
Diameter (Metric) 50 mm