Learn More
Black PN (~80%), TRC
Product Code. 30132546
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132546 10 g 10g
30134438 1 g 1g
30007428 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132546 Supplier Toronto Research Chemicals Supplier No. TRCB59200010G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2519-30-4
Molecular Formula C28 H17 N5 O14 S4 . 4 Na
Molecular Weight (g/mol) 867.68
Synonym 1,7-Naphthalenedisulfonic acid, 4-(acetylamino)-5-hydroxy-6-[[7-sulfo-4-[(4-sulfophenyl)azo]-1-naphthalenyl]azo]-, tetrasodium salt (9CI), Brilliant Black BN (6CI), C.I. Food Black 1 (7CI), C.I. Food Black 1, tetrasodium salt (8CI), 1743 Black, Black PN, Blue Black BN, Brilliant Acid Black BN Extra Pure A, Brilliant Acid Black BNA Export, Brilliant Black 1, Brilliant Black 80, Brilliant Black A, Brilliant Black N, Brilliant Black NAF, Brilliant Black NFQ, Brilliant Black PN, C.I. 28440, Certicol Black PNW, Cilefa Black B, E 151, Edicol Supra Black BN, Food Black 1, Hexacol Black PN, L Black 8000, Melan Black, Tetrasodium 2-[4-(p-sulfophenylazo)-7-sulfo-1-naphthylazo]-8-acetamido-1-naphthol-3,5-disulfonate, Xylene Black F
IUPAC Name tetrasodium;4-acetamido-5-hydroxy-6-[(E)-[7-sulfonato-4-[(Z)-(4-sulfonatophenyl)diazenyl]naphthalen-1-yl]diazenyl]naphthalene-1,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC(=O)Nc1ccc(c2cc(c(N=Nc3ccc(N=Nc4ccc(cc4)S(=O)(=O)[O-])c5ccc(cc35)S(=O)(=O)[O-])c(O)c12)S(=O)(=O)[O-])S(=O)(=O)[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C28H21N5O14S4.4Na/c1-14(34)29-23-10-11-24(50(42,43)44)20-13-25(51(45,46)47)27(28(35)26(20)23)33-32-22-9-8-21(18-7-6-17(12-19(18)22)49(39,40)41)31-30-15-2-4-16(5-3-15)48(36,37)38;;;;/h2-13,35H,1H3,(H,29,34)(H,36,37,38)(H,39,40,41)(H,42,43,44)(H,45,46,47);;;;/q;4*+1/p-4
Formula Weight 866.9246
Chemical Name or Material Brilliant Black BN