Learn More
Gravis Black Phenolic Resin Screw Caps
Product Code. 11733863
Click to view available options
Liner Material:
Butyl-PTFE
Polyethylene Foam
Polyethylene-coated Chipboard
Polyvinyl
Rubber
Thread Size (Metric):
18mm
18mm Polyex
18mm Teflon™
28mm
28mm PVDC
28mm Polyex
30mm
30mm Polyex
30mm Teflon™
33mm AS
35mm Polyex
35mm Teflon™
38mm TS
40mm
40mm Polyex
40mm Teflon™
48mm AS
Unit Size:
Pack of 100
Product Code. 11733863
Brand: Gravis P09030S100
Only null left
Add to basket
Edge
Add to basket
Providing secure closure for a variety of bottle neck sizes. Gravis Black Phenloic Resin Screw Caps are durable and resilient.
  • Polyethylene-coated chipboard liners temperature resistant up to 80°C
  • Rubber liners fully autoclavable
  • Polyethylene foam liners temperature resistant up to 60°C
  • Butyl-PTFE liners fully autoclavable
Specifications
Color Black
Material Phenolic Resin
No. per Pack 100Pack
Quantity 100 Pack
Thread Size (Metric) 30mm Polyex
Liner Material Polyethylene Foam
Product Type Cap