Learn More
Macherey-Nagel™ Black Filter Paper MN 220
Product Code. 10042651 Shop All Macherey-Nagel Products
Change view
Click to view available options
70 mm diameter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10042651 70 mm diameter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10042651 Supplier Macherey-Nagel™ Supplier No. 409007
Add to basket
Add to basket
Used to identify small quantities of light precipitates.
  • Dyed black with a sulfur dye
  • Used to identify small quantities of light precipitates such as the detection of fluorine or silicone
Specifications
Basis Weight 85 g/m2
Filtration Speed 45 sec.
Grade MN 220
Quantity 100/Pk.
Thickness (Metric) 0.17 mm
Diameter (Metric) 70 mm